Triphenylsulphonium chloride 95%

Lieferant: Apollo Scientific
Achtung

OR40449-1G OR40449-5G
APOSOR40449-1GEA 0 EUR
APOSOR40449-1G APOSOR40449-5G
Triphenylsulphonium chloride 95%
Triphenylsulphonium chloride
Formel: (C₆H₅)₃SCl
Lagertemperatur: Raumtemperatur
MDL: MFCD00031716
CAS-Nummer: 4270-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR