2-Methoxy-4-nitrophenol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR40450-100G
APOSOR40450-100GEA
0
EUR
APOSOR40450-100G
2-Methoxy-4-nitrophenol 98%
2-Methoxy-4-nitrophenol
Formel:
C₇H₇NO₄ Schmelzpunkt: 99…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012143 CAS-Nummer: 3251-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...