Bis(2-methyl-2-propanyl)-[(Z)-1H-pyrazol-1-ylmethylyliden]biscarbamat 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR40453-25G
OR40453-5G
APOSOR40453-25GEA
0
EUR
APOSOR40453-25G
APOSOR40453-5G
Bis(2-methyl-2-propanyl)-[(Z)-1H-pyrazol-1-ylmethylyliden]biscarbamat 98%
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate
Formel:
C₁₄H₂₂N₄O₄ Schmelzpunkt: 86…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683427 CAS-Nummer: 152120-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...