8-hydroxyquinoline-2-carboxylic acid 98%

Lieferant: Apollo Scientific

Synonyme: 8-Hydroxyquinoline-2-carboxylic acid

OR40470-1G
APOSOR40470-1GEA 0 EUR
APOSOR40470-1G
8-hydroxyquinoline-2-carboxylic acid 98%
8-hydroxyquinoline-2-carboxylic acid
Formel: C₁₀H₇NO₃
Molecular Weight: 189,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00168972
CAS-Nummer: 1571-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR