Tetrachlor-o-benzochinon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Tetrachlor-1,2-benzochinon, 3,4,5,6-Tetrachlor-1,2-benzochinon, o-Chloranil, 2-Chloranil
OR40496-5G
APOSOR40496-5GEA
195
EUR
APOSOR40496-5G
Tetrachlor-o-benzochinon
Tetrachlor-o-benzochinon
Formel:
C₆Cl₄O₂ Molecular Weight: 245,88 g/mol Schmelzpunkt: 126…129 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001646 CAS-Nummer: 2435-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...