2-tert-Butylbenzoic acid

Lieferant: Apollo Scientific
Achtung

OR40497-1G
APOSOR40497-1GEA 208 EUR
APOSOR40497-1G
2-tert-Butylbenzoic acid
2-tert-Butylbenzoic acid
Formel: C₁₁H₁₄O₂
Schmelzpunkt: 68…69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09258829
CAS-Nummer: 1077-58-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR