Methyl-2-methyl-1,3-benzoxazole-7-carboxylate

Lieferant: Apollo Scientific

OR40499-1G
APOSOR40499-1GEA 290 EUR
APOSOR40499-1G
Methyl-2-methyl-1,3-benzoxazole-7-carboxylate
Methyl-2-methyl-1,3-benzoxazole-7-carboxylate
Formel: C₁₀H₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11042247
CAS-Nummer: 1234847-45-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR