3-Chloro-4-(methylsulphonyl)thiophene-2-carbonyl chloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-5-(chlorocarbonyl)thien-3-yl methyl sulphone, 3-Chloro-2-(chlorocarbonyl)-4-(methylsulphonyl)thiophene

OR4050-1G
APOSOR4050-1GEA 380 EUR
APOSOR4050-1G
3-Chloro-4-(methylsulphonyl)thiophene-2-carbonyl chloride
3-Chloro-4-(methylsulphonyl)thiophene-2-carbonyl chloride
Formel: C₆H₄Cl₂O₃S₂
Molecular Weight: 259,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00174300
CAS-Nummer: 175201-87-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR