Ethyl-7-chlorimidazo[1,2-a]pyridin-2-carboxylat
Lieferant: Apollo Scientific
|
Achtung
|
OR40503-1G
APOSOR40503-1GEA
154
EUR
APOSOR40503-1G
Ethyl-7-chlorimidazo[1,2-a]pyridin-2-carboxylat
Ethyl-7-chlorimidazo[1,2-a]pyridin-2-carboxylat
Formel:
C₁₀H₉BrN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11505035 CAS-Nummer: 1204501-38-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...