Ethyl-4-methylthiazole-5-carboxylate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Methylthiazole-5-carboxylic acid ethyl ester, Ethyl 4-methylthiazole-5-carboxylate
OR40519-25G
APOSOR40519-25GEA
56.7
EUR
APOSOR40519-25G
Ethyl-4-methylthiazole-5-carboxylate 98%
Ethyl-4-methylthiazole-5-carboxylate
Formel:
C₇H₉NO₂S Molecular Weight: 171.214 g/mol Siedepunkt: 215…233 °C Schmelzpunkt: 27…28 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00626873 CAS-Nummer: 20582-55-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...