Ethyl-2-amino-4-methyl-5-oxazolcarboxylat 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-amino-4-methyloxazole-5-carboxylate

OR40536-1G
APOSOR40536-1GEA 72.5 EUR
APOSOR40536-1G
Ethyl-2-amino-4-methyl-5-oxazolcarboxylat 95+%
Ethyl-2-amino-4-methyl-5-oxazolcarboxylat
Formel: C₇H₁₀N₂O₃
Molecular Weight: 170,1659 g/mol
Schmelzpunkt: 215…217 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09028430
CAS-Nummer: 79221-15-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR