7-Bromo-1-benzothiophene-2-carboxylic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Bromobenzo[b]thiophene-2-carboxylic acid

OR40543-5G
APOSOR40543-5GEA 98.6 EUR
APOSOR40543-5G
7-Bromo-1-benzothiophene-2-carboxylic acid 98%
7-Bromo-1-benzothiophene-2-carboxylic acid
Formel: C₉H₅BrO₂S
Molecular Weight: 257.11 g/mol
Schmelzpunkt: >275 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12032270
CAS-Nummer: 19075-59-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR