5-Methyl-2-furohydrazide

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Methylfuran-2-carboxylic acid hydrazide, 5-Methylfuran-2-carbohydrazide

OR40552-1G
APOSOR40552-1GEA 163 EUR
APOSOR40552-1G
5-Methyl-2-furohydrazide
5-Methyl-2-furohydrazide
Formel: C₆H₈N₂O₂
Molecular Weight: 140,14 g/mol
Schmelzpunkt: 61…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01365944
CAS-Nummer: 20842-19-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR