4-Hydroxy-o-toluylsäure 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Hydroxy-2-methylbenzoic acid, 4-Hydroxy-o-toluic acid
OR40557-25G
APOSOR40557-25GEA
125
EUR
APOSOR40557-25G
4-Hydroxy-o-toluylsäure 97%
4-Hydroxy-o-toluylsäure
Formel:
C₈H₈O₃ Molecular Weight: 152,15 g/mol Schmelzpunkt: 177…181 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02182261 CAS-Nummer: 578-39-2 EINECS: 209-422-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...