Ethyl 2-(4-Oxocyclohexyl)acetate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2-(4-oxocyclohexyl)acetate, 2-(4-Oxocyclohexyl)acetic acid Ethyl ester
OR40563-1G
APOSOR40563-1GEA
21.9
EUR
APOSOR40563-1G
Ethyl 2-(4-Oxocyclohexyl)acetate 95%
Ethyl 2-(4-Oxocyclohexyl)acetate
Formel:
C₁₀H₁₆O₃ Molecular Weight: 184,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11045783 CAS-Nummer: 58012-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...