2-Bromo-6-chlorobenzyl bromide 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-Bromo-2-(bromomethyl)-3-chlorobenzene, 2-Chloro-α,6-dibromotoluene
OR40565-5G
APOSOR40565-5GEA
134
EUR
APOSOR40565-5G
2-Bromo-6-chlorobenzyl bromide 97%
2-Bromo-6-chlorobenzyl bromide
Formel:
C₇H₅Br₂Cl Molecular Weight: 284.376 g/mol Schmelzpunkt: 58…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038394 CAS-Nummer: 75002-98-1 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...