tert-Butyl 4-bromobutanoate 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 4-bromobutyrate

OR40573-25G
APOSOR40573-25GEA 197 EUR
APOSOR40573-25G
tert-Butyl 4-bromobutanoate 96%
tert-Butyl 4-bromobutanoate
Formel: C₈H₁₅BrO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02179416
CAS-Nummer: 110661-91-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR