4-[(4,6-Dimethoxypyrimidin-2-yl)methoxy]benzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[(4-Carboxyphenoxy)methyl]-4,6-dimethoxypyrimidine, 4-[(4,6-Dimethoxy-1,3-diazin-2-yl)methoxy]benzoic acid

OR4058-50MG
APOSOR4058-50MGEA 42.2 EUR
APOSOR4058-50MG
4-[(4,6-Dimethoxypyrimidin-2-yl)methoxy]benzoic acid
4-[(4,6-Dimethoxypyrimidin-2-yl)methoxy]benzoic acid
Formel: C₁₄H₁₄N₂O₅
Molecular Weight: 290.271 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01567897
CAS-Nummer: 402497-58-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR