4'-Amino-3'-nitroacetanilide

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Acetamido-2-nitroaniline, N-(4-Amino-3-nitrophenyl)acetamide

OR40581-250MG
APOSOR40581-250MGEA 37.7 EUR
APOSOR40581-250MG
4'-Amino-3'-nitroacetanilide
4'-Amino-3'-nitroacetanilide
Formel: C₈H₉N₃O₃
Molecular Weight: 195.175 g/mol
Schmelzpunkt: 224…228 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00035890
CAS-Nummer: 6086-29-9
EINECS: 228-019-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR