(3R)-3-Aminopentanoic acid hydrochloride

Lieferant: Apollo Scientific
Achtung

OR40585-1G OR40585-250MG
APOSOR40585-1GEA 0 EUR
APOSOR40585-1G APOSOR40585-250MG
(3R)-3-Aminopentanoic acid hydrochloride
(3R)-3-Aminopentanoic acid hydrochloride
Formel: C₅H₁₁NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD08702981
CAS-Nummer: 952650-02-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR