4-[2-(Boc-amino)ethyl]aniline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 4-aminophenethylcarbamate, 4-(2-Aminoethyl)aniline, 4-BOC protected
OR40588-1G
APOSOR40588-1GEA
21.9
EUR
APOSOR40588-1G
4-[2-(Boc-amino)ethyl]aniline
4-[2-(Boc-amino)ethyl]aniline
Formel:
C₁₃H₂₀N₂O₂ Molecular Weight: 236,315 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD05663961 CAS-Nummer: 94838-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...