Ethyl 4-chloro-1,3-dimethylpyrazolo[3,4-b]pyridine-5-carboxylate

Lieferant: Apollo Scientific

OR4060-1G
APOSOR4060-1GEA 87 EUR
APOSOR4060-1G
Ethyl 4-chloro-1,3-dimethylpyrazolo[3,4-b]pyridine-5-carboxylate
Ethyl 4-chloro-1,3-dimethylpyrazolo[3,4-b]pyridine-5-carboxylate
Formel: C₁₁H₁₂ClN₃O₂
Molecular Weight: 253,685 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00067954
CAS-Nummer: 20481-15-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR