6-Chloro-3-[(dimethylamino)methyl]imidazo[1,2-a]pyridine hydrochloride
Lieferant: Apollo Scientific
OR40602-1G
APOSOR40602-1GEA
232
EUR
APOSOR40602-1G
6-Chloro-3-[(dimethylamino)methyl]imidazo[1,2-a]pyridine hydrochloride
6-Chloro-3-[(dimethylamino)methyl]imidazo[1,2-a]pyridine hydrochloride
Formel:
C₁₀H₁₂ClN₃·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD12756250 CAS-Nummer: 1227954-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...