3-Amino-5-methoxyindan-1-one hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-2,3-dihydro-5-methoxy-1H-inden-1-one hydrochloride
OR40603-250MG
APOSOR40603-250MGEA
112
EUR
APOSOR40603-250MG
3-Amino-5-methoxyindan-1-one hydrochloride
3-Amino-5-methoxyindan-1-one hydrochloride
Formel:
C₁₀H₁₁NO₂·ClH Molecular Weight: 213,661 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01210167 CAS-Nummer: 148502-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...