Methyl 3-acetylbenzoate

Lieferant: Apollo Scientific
Achtung

OR40606-1G OR40606-5G OR40606-10G
APOSOR40606-1GEA 0 EUR
APOSOR40606-1G APOSOR40606-5G APOSOR40606-10G
Methyl 3-acetylbenzoate
Methyl 3-acetylbenzoate
Formel: C₁₀H₁₀O₃
Schmelzpunkt: 43…46 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09475799
CAS-Nummer: 21860-07-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR