4-Chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-1,3-dimethyl-7-azaindazole-5-carboxylic acid, 5-Carboxy-4-chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine

OR4061-1G
APOSOR4061-1GEA 58 EUR
APOSOR4061-1G
4-Chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid 97%
4-Chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid
Formel: C₉H₈ClN₃O₂
Molecular Weight: 225,632 g/mol
Schmelzpunkt: 269 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00067959
CAS-Nummer: 175201-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR