4H-Amino-1,3-dihydroisothiochromen-1,1-dioxide hydrochloride 95+%

Lieferant: Apollo Scientific

Synonyme: 4-Aminoisothiochroman 2,2-dioxide hydrochloride

OR40611-250MG
APOSOR40611-250MGEA 201 EUR
APOSOR40611-250MG
4H-Amino-1,3-dihydroisothiochromen-1,1-dioxide hydrochloride 95+%
4H-Amino-1,3-dihydroisothiochromen-1,1-dioxide hydrochloride
Formel: C₉H₁₂ClNO₂S
Molecular Weight: 233,72 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11616514
CAS-Nummer: 1187830-61-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR