5-Pyridin-2-ylthiophene-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR4063-1G
APOSOR4063-1GEA
92.8
EUR
APOSOR4063-1G
5-Pyridin-2-ylthiophene-2-carboxylic acid
5-Pyridin-2-ylthiophene-2-carboxylic acid
Formel:
C₁₀H₇NO₂S Molecular Weight: 205,24 g/mol Schmelzpunkt: 231…233 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00052899 CAS-Nummer: 119082-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...