trans-4-Hydroxycyclohexancarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR40634-25G
OR40634-5G
OR40634-100G
APOSOR40634-25GEA
0
EUR
APOSOR40634-25G
APOSOR40634-5G
APOSOR40634-100G
trans-4-Hydroxycyclohexancarbonsäure
trans-4-Hydroxycyclohexanecarboxylic acid
Formel:
C₇H₁₂O₃ Schmelzpunkt: 145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093477 CAS-Nummer: 3685-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...