3-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)propionaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Phthalimid-1-yl)propanal, 3-(1,3-Dioxoisoindolin-2-yl)propanal

OR40638-1G
APOSOR40638-1GEA 23.2 EUR
APOSOR40638-1G
3-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)propionaldehyde 95%
3-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)propionaldehyde
Formel: C₁₁H₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01704466
CAS-Nummer: 2436-29-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR