5-{[2-(Methylamino)-5-nitrophenyl]amino}-5-oxopentanoic acid 97%

Lieferant: Apollo Scientific

Synonyme: 5-((2-(Methylamino)-5-nitrophenyl)amino)-5-oxopentanoic acid

OR40639-1G
APOSOR40639-1GEA 81.2 EUR
APOSOR40639-1G
5-{[2-(Methylamino)-5-nitrophenyl]amino}-5-oxopentanoic acid 97%
5-{[2-(Methylamino)-5-nitrophenyl]amino}-5-oxopentanoic acid
Formel: C₁₂H₁₅N₃O₅
Molecular Weight: 281,265 g/mol
Schmelzpunkt: 170,5…171.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09840668
CAS-Nummer: 91644-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR