2-Amino-3-furonitrile 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-3-cyanofuran, 2-Aminofuran-3-carbonitrile
OR40643-250MG
APOSOR40643-250MGEA
236
EUR
APOSOR40643-250MG
2-Amino-3-furonitrile 95+%
2-Amino-3-furonitrile
Formel:
C₅H₄N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09834958 CAS-Nummer: 139370-56-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...