(R)-(-)-1-Boc-3-(aminomethyl)pyrrolidine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3R)-3-(Aminomethyl)pyrrolidine, N1-BOC protected, (R)-tert-Butyl 3-(aminomethyl)pyrrolidine-1-carboxylate
OR40644-1G
APOSOR40644-1GEA
81.2
EUR
APOSOR40644-1G
(R)-(-)-1-Boc-3-(aminomethyl)pyrrolidine 97%
(R)-(-)-1-Boc-3-(aminomethyl)pyrrolidine
Formel:
C₁₀H₂₀N₂O₂ Molecular Weight: 200,282 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03844734 CAS-Nummer: 199174-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...