4-iodobenzene-1,2-diamine 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR40649-5G
APOSOR40649-5GEA
111
EUR
APOSOR40649-5G
4-iodobenzene-1,2-diamine 97%
4-iodobenzene-1,2-diamine
Formel:
C₆H₇IN₂ Schmelzpunkt: 73…77 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08669466 CAS-Nummer: 21304-38-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...