3-Amino-6-brom-2-pyrazincarbonsäure

Lieferant: Apollo Scientific
Achtung

OR40653-1G
APOSOR40653-1GEA 21.9 EUR
APOSOR40653-1G
3-Amino-6-brom-2-pyrazincarbonsäure
3-Amino-6-bromopyrazine-2-carboxylic acid
Formel: C₅H₄BrN₃O₂
Molecular Weight: 218.01 g/mol
Schmelzpunkt: 178…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08275681
CAS-Nummer: 486424-37-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR