3,5-Dimethyl-1H-pyrazol-4-boronsäure Hydrochlorid

Lieferant: Apollo Scientific
Achtung

OR40658-250MG
APOSOR40658-250MGEA 125 EUR
APOSOR40658-250MG
3,5-Dimethyl-1H-pyrazol-4-boronsäure Hydrochlorid
3,5-Dimethyl-1H-pyrazol-4-boronsäure Hydrochlorid
Formel: C₅H₉BN₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD11616525
CAS-Nummer: 1162262-39-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR