2-Bromo-5-iodobenzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
α,2-Dibromo-5-iodotoluene, 1-Bromo-2-(bromomethyl)-4-iodobenzene
OR40661-1G
OR40661-250MG
OR40661-5G
APOSOR40661-1GEA
140
EUR
APOSOR40661-1G
APOSOR40661-250MG
APOSOR40661-5G
2-Bromo-5-iodobenzyl bromide
2-Bromo-5-iodobenzyl bromide
Formel:
C₇H₅Br₂I Lagertemperatur: Raumtemperatur |
MDL:
MFCD15527653 CAS-Nummer: 289617-98-7 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...