2-Bromo-5-iodobenzyl bromide

Lieferant: Apollo Scientific
Gefahr

Synonyme: α,2-Dibromo-5-iodotoluene, 1-Bromo-2-(bromomethyl)-4-iodobenzene

OR40661-1G OR40661-250MG OR40661-5G
APOSOR40661-1GEA 140 EUR
APOSOR40661-1G APOSOR40661-250MG APOSOR40661-5G
2-Bromo-5-iodobenzyl bromide
2-Bromo-5-iodobenzyl bromide
Formel: C₇H₅Br₂I
Lagertemperatur: Raumtemperatur
MDL: MFCD15527653
CAS-Nummer: 289617-98-7
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR