3-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid monohydrochloride monohydrate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Carboxy-3-chloroimidazo[1,2-a]pyridine monohydrochloride monohydrate

OR40684-250MG
APOSOR40684-250MGEA 65.4 EUR
APOSOR40684-250MG
3-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid monohydrochloride monohydrate
3-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid monohydrochloride monohydrate
Formel: C₈H₅ClN₂O₂·ClH·H₂O
Molecular Weight: 251,067 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27995551
CAS-Nummer: 2022965-72-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR