6-Chlorisatin 97+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Coroindoline-2,3-dione
OR40689-100G
OR40689-25G
APOSOR40689-100GEA
249
EUR
APOSOR40689-100G
APOSOR40689-25G
6-Chlorisatin 97+%
6-Chlorisatin
Formel:
C₈H₄ClNO₂ Schmelzpunkt: 262 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00086347 CAS-Nummer: 6341-92-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...