4-Brom-o-anisaldehyd 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-methoxybenzaldehyde

OR40709-25G
APOSOR40709-25GEA 74.1 EUR
APOSOR40709-25G
4-Brom-o-anisaldehyd 98%
4-Brom-o-anisaldehyd
Formel: C₈H₇BrO₂
Molecular Weight: 215.05 g/mol
Schmelzpunkt: 67…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08276823
CAS-Nummer: 43192-33-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR