2-(Cyanomethyl)benzeneboronic acid pinacol ester 97%

Lieferant: Apollo Scientific
Achtung

OR4072-250MG
APOSOR4072-250MGEA 27.7 EUR
APOSOR4072-250MG
2-(Cyanomethyl)benzeneboronic acid pinacol ester 97%
2-(Cyanomethyl)benzeneboronic acid pinacol ester
Formel: C₁₄H₁₈BNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02179457
CAS-Nummer: 325141-71-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR