4-(Furfurylcarbamoyl)phenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-((Furan-2-ylmethyl)carbamoyl)phenyl)boronic acid, 4-[(Fur-2-ylmethyl)carbamoyl]benzeneboronic acid

OR4073-1G
APOSOR4073-1GEA 23.2 EUR
APOSOR4073-1G
4-(Furfurylcarbamoyl)phenylboronsäure 97%
4-(Furfurylcarbamoyl)phenylboronsäure
Formel: C₁₂H₁₂BNO₄
Schmelzpunkt: 134…140 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04115672
CAS-Nummer: 850568-18-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR