3-Chloro-5-(chloromethyl)pyridine hydrochloride 97%

Lieferant: Apollo Scientific
Gefahr

OR40752-1G
APOSOR40752-1GEA 364 EUR
APOSOR40752-1G
3-Chloro-5-(chloromethyl)pyridine hydrochloride 97%
3-Chloro-5-(chloromethyl)pyridine hydrochloride
Formel: C₆H₅Cl₂N·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD16037340
CAS-Nummer: 847610-86-0
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR