2-Bromo-4-cyclopropylaniline 96%

Lieferant: Apollo Scientific
Achtung

OR40777-1G OR40777-250MG OR40777-5G
APOSOR40777-1GEA 493 EUR
APOSOR40777-1G APOSOR40777-250MG APOSOR40777-5G
2-Bromo-4-cyclopropylaniline 96%
2-Bromo-4-cyclopropylaniline
Formel: C₉H₁₀BrN
Molecular Weight: 212.086 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD21099997
CAS-Nummer: 1353857-13-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR