2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Lieferant: Apollo Scientific
OR4078-1G
OR4078-250MG
APOSOR4078-1GEA
85.7
EUR
APOSOR4078-1G
APOSOR4078-250MG
2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Formel:
C₉H₉N₃O₂ Molecular Weight: 191.19 g/mol Schmelzpunkt: 245…247 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067901 CAS-Nummer: 175201-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...