tert-Butyl [3-(aminomethyl)oxetan-3-yl]carbamate 95%

Lieferant: Apollo Scientific

OR40813-250MG
APOSOR40813-250MGEA 841 EUR
APOSOR40813-250MG
tert-Butyl [3-(aminomethyl)oxetan-3-yl]carbamate 95%
tert-Butyl [3-(aminomethyl)oxetan-3-yl]carbamate
Formel: C₉H₁₈N₂O₃
Molecular Weight: 202,251 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29042833
CAS-Nummer: 1802048-96-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR