(4-Prop-2-ynoxyphenyl)boronic acid, pinacol ester 95%

Lieferant: Apollo Scientific
Achtung

OR40822-1G OR40822-250MG OR40822-5G
APOSOR40822-1GEA 319 EUR
APOSOR40822-1G APOSOR40822-250MG APOSOR40822-5G
(4-Prop-2-ynoxyphenyl)boronic acid, pinacol ester 95%
(4-Prop-2-ynoxyphenyl)boronic acid, pinacol ester
Formel: C₁₅H₁₉BO₃
Lagertemperatur: Kühlschrank
MDL: MFCD32177102
CAS-Nummer: 2280980-72-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR