3-Chlorbenzoylacetonitril
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(3-Chlorphenyl)-3-oxopropionsäurenitril, 3-(3-Chlorphenyl)-3-ketopropionsäurenitril
OR4086-1G
APOSOR4086-1GEA
21.9
EUR
APOSOR4086-1G
3-Chlorbenzoylacetonitril
3-Chlorbenzoylacetonitril
Formel:
C₉H₆ClNO Molecular Weight: 179,61 g/mol Schmelzpunkt: 82…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067891 CAS-Nummer: 21667-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...