2-(Methylthio)thiophen

Lieferant: Apollo Scientific
Achtung

OR4091-10G OR4091-5G OR4091-1G
APOSOR4091-10GEA 0 EUR
APOSOR4091-10G APOSOR4091-5G APOSOR4091-1G
2-(Methylthio)thiophen
2-(Methylthio)thiophene
Formel: C₅H₆S₂
Siedepunkt: 82…84 °C/15mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD00052382
CAS-Nummer: 5780-36-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR